Skip to Content
Merck
All Photos(1)

Key Documents

307858

Sigma-Aldrich

Scandium(III) chloride hydrate

99.9%

Synonym(s):

Scandium trichloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$232.00

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$232.00

About This Item

Linear Formula:
ScCl3·xH2O
CAS Number:
Molecular Weight:
151.31 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.9%

form

solid

composition

Degree of hydration, 5-7

reaction suitability

core: scandium
reagent type: catalyst

SMILES string

[H]O[H].Cl[Sc](Cl)Cl

InChI

1S/3ClH.H2O.Sc/h3*1H;1H2;/q;;;;+3/p-3

InChI key

UJMNPKCPAGGCBI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Prins cyclization reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Po-Jung Jimmy Huang et al.
Analytical chemistry, 87(13), 6890-6895 (2015-06-11)
Mercury is a highly toxic heavy metal, and detection of Hg(2+) by biosensors has attracted extensive research interest in the past decade. In particular, a number of DNA-based sensing strategies have been developed. Well-known examples include thymine-Hg(2+) interactions and Hg(2+)-activated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service