Skip to Content
Merck
All Photos(3)

Key Documents

307076

Sigma-Aldrich

4′-Methylacetanilide

99%

Synonym(s):

p-Acetotoluidide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4CH3
CAS Number:
Molecular Weight:
149.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

307 °C (lit.)

mp

149-151 °C (lit.)

SMILES string

CC(=O)Nc1ccc(C)cc1

InChI

1S/C9H11NO/c1-7-3-5-9(6-4-7)10-8(2)11/h3-6H,1-2H3,(H,10,11)

InChI key

YICAMJWHIUMFDI-UHFFFAOYSA-N

Application

4′-Methylacetanilide was used in the synthesis of stable crystalline phosphorus ylides[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of stable phosphorus ylides derived from triphenylphosphine, dialkyl acetylenedicarboxylates, and an NH-Acid.
Phosphorus, Sulfur, and Silicon and the Related Elements, 181(4), 865-877 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service