Skip to Content
Merck
All Photos(1)

Key Documents

30695

Sigma-Aldrich

Chloro-decyl-dimethylsilane

≥97.0% (GC)

Synonym(s):

Decyl-dimethyl-chlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27ClSi
CAS Number:
Molecular Weight:
234.88
Beilstein:
2322585
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.441

bp

98 °C/2 mmHg (lit.)

density

0.866 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCC[Si](C)(C)Cl

InChI

1S/C12H27ClSi/c1-4-5-6-7-8-9-10-11-12-14(2,3)13/h4-12H2,1-3H3

InChI key

ZLZGHBNDPINFKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

278.6 °F - closed cup

Flash Point(C)

137 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service