Skip to Content
Merck
All Photos(1)

Key Documents

306134

Sigma-Aldrich

Tridodecylamine

technical grade, 85%

Synonym(s):

Hydrogen ionophore I

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N
CAS Number:
Molecular Weight:
521.99
Beilstein:
1801669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

vapor density

>1 (vs air)

Assay

85%

refractive index

n20/D 1.4578 (lit.)

bp

220-228 °C/0.03 mmHg (lit.)

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C36H75N/c1-4-7-10-13-16-19-22-25-28-31-34-37(35-32-29-26-23-20-17-14-11-8-5-2)36-33-30-27-24-21-18-15-12-9-6-3/h4-36H2,1-3H3

InChI key

SWZDQOUHBYYPJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

374.0 °F

Flash Point(C)

190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G T McEwan et al.
Journal of medical engineering & technology, 14(1), 16-20 (1990-01-01)
Incorporation of a ligand into polyvinylchloride allows the production of ion selective electrodes that are soft-bodied and disposable. With features make them especially suitable for clinical, particularly gastroenterological, investigations. We report here on the construction of a combined reference and
M O'Dowd et al.
Journal of biomedical engineering, 10(2), 165-170 (1988-04-01)
Frequent measurement of pH and PCO2 levels of fetal blood during labour provides an insight into the presence and nature of fetal asphyxia, effectiveness of in utero resuscitation and the need for operative delivery. A pH measuring system utilizing a
Jacobine J H A van Hest et al.
Nanoscale research letters, 11(1), 261-261 (2016-05-23)
Lanthanide ions are promising for the labeling of silica nanoparticles with a specific luminescent fingerprint due to their sharp line emission at characteristic wavelengths. With the increasing use of silica nanoparticles in consumer products, it is important to label silica
J M Rawlings et al.
Gut, 26(2), 203-207 (1985-02-01)
Plastic electrodes have been developed for measuring pH in the human gastrointestinal tract. The electrodes have a plastic hydrogen ion sensitive membrane sealed to a length of fluid filled PVC tubing. Two recently developed hydrogen ion sensitive ligands have been
Shiho Kondo et al.
American journal of physiology. Gastrointestinal and liver physiology, 309(4), G260-G269 (2015-06-20)
Although cystic fibrosis is rare in Japanese, measurement of sweat Cl(-) has suggested mild dysfunction of cystic fibrosis transmembrane conductance regulator (CFTR) in some patients with chronic pancreatitis. In the present study, we have investigated the association of CFTR variants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service