Skip to Content
Merck
All Photos(1)

Key Documents

305626

Sigma-Aldrich

Hexylphosphonic dichloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5P(O)Cl2
CAS Number:
Molecular Weight:
203.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

114-121 °C/16 mmHg (lit.)

density

1.185 g/mL at 25 °C (lit.)

functional group

phosphine oxide

SMILES string

CCCCCCP(Cl)(Cl)=O

InChI

1S/C6H13Cl2OP/c1-2-3-4-5-6-10(7,8)9/h2-6H2,1H3

InChI key

GZXBQDURAKBNAZ-UHFFFAOYSA-N

Application

Hexylphosphonic dichloride was used in preparation of:
  • hexylphosphonic acid via reaction with water[1]
  • phosphonate esters[2]
  • methyl 4-methylumbelliferyl hexylphosphonate[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of the shape evolution of CdSe nanocrystals.
Peng ZA and Peng X.
Journal of the American Chemical Society, 123(7), 1389-1395 (2001)
Active-site titration analysis of surface influences on immobilized< i> Candida antarctica</i> lipase B activity.
Laszlo JA, et al.
Journal of Molecular Catalysis. B, Enzymatic, 69(1), 60-65 (2011)
Alessandra Mezzetti et al.
Chemistry & biology, 12(4), 427-437 (2005-04-27)
Synthetic chemists often exploit the high enantioselectivity of lipases to prepare pure enantiomers of primary alcohols, but the molecular basis for this enantioselectivity is unknown. The crystal structures of two phosphonate transition-state analogs bound to Burkholderia cepacia lipase reveal this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service