Skip to Content
Merck
All Photos(2)

Key Documents

305413

Sigma-Aldrich

Poly(ethylene glycol) distearate

average MN 930, cross-linking reagent click chemistry

Synonym(s):

Polyethylene glycol, Polyoxyethylene bis(stearate)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$220.00

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$220.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) distearate, average Mn ~930

mol wt

average Mn ~930

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

mp

35-37 °C

transition temp

softening point 35-37 °C

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

OCCO.CCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C17H34O2.C2H6O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19;3-1-2-4/h2-16H2,1H3,(H,18,19);3-4H,1-2H2

InChI key

QYJQHDWGURXGNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Optimization of lipase-catalyzed synthesis of polyethylene glycol stearate in a solvent-free system: This research discusses the enzymatic synthesis of polyethylene glycol stearate, widely used in the pharmaceutical and cosmetic industries. The study emphasizes optimizing production conditions to enhance yield and efficiency (Nhivekar & Rathod, 2019).
  • Application of the response surface methodology for modeling demulsification of crude oil emulsion using a demulsifier: This article explores the use of polyethylene glycol distearate in demulsifying crude oil emulsions, highlighting its effectiveness and application in industrial processes (Roshan et al., 2018).
  • Synthesis and nanoencapsulation of poly (ethylene glycol)-distearates phase change materials for latent heat storage and release: This research focuses on the application of polyethylene glycol distearate in creating phase change materials for energy storage, demonstrating its utility in enhancing thermal energy storage capabilities (Kumar et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service