Skip to Content
Merck
All Photos(1)

Key Documents

305081

Sigma-Aldrich

4-Methoxy-2-quinolinecarboxylic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

197 °C (dec.) (lit.)

SMILES string

COc1cc(nc2ccccc12)C(O)=O

InChI

1S/C11H9NO3/c1-15-10-6-9(11(13)14)12-8-5-3-2-4-7(8)10/h2-6H,1H3,(H,13,14)

InChI key

AVBKMSSLAIKOGM-UHFFFAOYSA-N

Gene Information

General description

The redox chemistry of the ligand 4-methoxy-2-quinolinecarboxylic acid, its monoanion and its complexes with manganese(II) and manganese(III) was studied.[1]

Application

4-Methoxy-2-quinolinecarboxylic acid was used in preparation of novel oxorhenium(V) complexes incorporating quinoline and isoquinoline carboxylic acid derivatives[2] and 2,3-diaminopyridinium 4-methoxyquinoline-2-carboxylate.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manganese (II) and (III) complexes with 4-methoxy-2-quinolinecarboxylic acid: Mono-and binuclear in aprotic medium.
Bodini ME and Arancibia MV.
Polyhedron, 11(17), 2195-2202 (1992)
Barbara Machura et al.
Dalton transactions (Cambridge, England : 2003), 42(24), 8827-8837 (2013-05-04)
Six novel oxorhenium(V) complexes incorporating quinoline and isoquinoline carboxylic acid derivatives were prepared in good yields. Relying on the experimental conditions, compounds with two chelate ligands [ReOCl(iqc)2]·MeOH (1), [ReO(OMe)(iqc)2] (2), [ReO(OMe)(mqc)2] (3) and [ReO(OMe)(8-qc)2] (4) and compounds incorporating one bidentate
Kaliyaperumal Thanigaimani et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 12), o3444-o3445 (2013-03-12)
In the 4-meth-oxy-quinoline-2-carboxyl-ate anion of the title salt, C5H8N3(+)·C11H8NO3(-), the dihedral angle between the quinoline ring system and the carboxyl-ate group is 16.54 (15)°. In the crystal, the cations and anions are linked via N-H⋯O and N-H⋯N hydrogen bonds, forming a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service