Skip to Content
Merck
All Photos(2)

Key Documents

30471

Sigma-Aldrich

Davy-reagent p-tolyl

≥97.0%

Synonym(s):

2,4-Bis(p-tolylthio)-1,3,2,4-dithiadiphosphetane 2,4-disulfide, DR-T, Heimgartner reagent

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$371.00

$371.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$371.00

About This Item

Empirical Formula (Hill Notation):
C14H14P2S6
CAS Number:
Molecular Weight:
436.60
Beilstein:
7222715
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$371.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥97.0%

mp

209-213 °C (dec.)

SMILES string

Cc1ccc(SP2(=S)SP(=S)(Sc3ccc(C)cc3)S2)cc1

InChI

1S/C14H14P2S6/c1-11-3-7-13(8-4-11)19-15(17)21-16(18,22-15)20-14-9-5-12(2)6-10-14/h3-10H,1-2H3

InChI key

WMYQGZHFMUDZIS-UHFFFAOYSA-N

Other Notes

Reagent for the thiation of amides showing complementary selectivity compared with the Lawesson-reagent[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Wipf et al.
Helvetica Chimica Acta, 70, 1001-1001 (1987)
Sambit Kumar Mishra et al.
Proteins, 85(8), 1422-1434 (2017-04-07)
It is known that over half of the proteins encoded by most organisms function as oligomeric complexes. Oligomerization confers structural stability and dynamics changes in proteins. We investigate the effects of oligomerization on protein dynamics and its functional significance for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service