Skip to Content
Merck
All Photos(1)

Key Documents

304026

Sigma-Aldrich

Sodium cyclopentadienylide

Sodium cyclopentadienylide

2.4 M in THF

Synonym(s):

Cyclopentadienylsodium solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Na
CAS Number:
Molecular Weight:
88.08
Beilstein:
969542
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reagent type: ligand

concentration

2.4 M in THF

density

0.938 g/mL at 25 °C

SMILES string

[Na]C1C=CC=C1

InChI

1S/C5H5.Na/c1-2-4-5-3-1;/h1-5H;

InChI key

PDTZVKVFAJGNRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

It can also be used in the synthesis of:
  • Ruthenocene and fullerene molecular hybrid, Ru(η5- C60Me5)( η5-C5H5) (bucky ruthenocene).[1]
  • Allyl-substituted cyclopentadiene by palladium-catalyzed allylic alkylation.[2]
  • Bis(cyclopentadienyl)zirconium dichloride which is used as a catalyst in the synthesis of bis(indolyl)methanes and alkylation of heteroaryls.[3]
  • Cp-cobaltacycles from diphenylphosphinoalkynes and CoCl(PPh3)3.[4]
Sodium cyclopentadienylide is a useful reagent for the preparation of metallocenes[5][6] and cyclopentadienyl metal complexes.[7]

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and reactivity of bucky ruthenocene Ru(η5- C60Me5)( η5-C5H5).
Matsuo Y, et al.
Chemistry Letters (Jpn), 33(1), 68-69 (2003)
Journal of Organometallic Chemistry, 467, 75-75 (1994)
Journal of the Chemical Society. Chemical Communications, 2007-2007 (1994)
Bis (cyclopentadienyl) zirconium dichloride for alkylation of heteroaromatics and synthesis of bis (indolyl) methanes.
Kantam ML, et al.
Catalysis Letters, 98(2-3), 117-121 (2004)
J. Chem. Soc., Dalton Trans., 2109-2109 (1994)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service