Skip to Content
Merck
All Photos(1)

Key Documents

303429

Sigma-Aldrich

Citronellic acid

98%

Synonym(s):

3,7-Dimethyl-6-octenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

refractive index

n20/D 1.454 (lit.)

bp

121-122 °C/1 mmHg (lit.)

density

0.923 g/mL at 25 °C (lit.)

SMILES string

CC(CC\C=C(\C)C)CC(O)=O

InChI

1S/C10H18O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,9H,4,6-7H2,1-3H3,(H,11,12)

InChI key

GJWSUKYXUMVMGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citronellic acid, β-methyl branched acid, is a terepenoid.[1] It is a major component isolated from the leaf oils of Pelargonium species.[2]

Application

(R)-(+)-Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.3

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stereocontrolled synthesis of all of the possible stereoisomers of 3, 11-dimethylnonacosan-2-one and 29-hydroxy-3, 11-dimethylnonacosan-2-one: The female sex pheromone of the german cockroach.
Mori K, et al.
Tetrahedron, 37(7), 1329-1340 (1981)
Citronellic acid: A major component in two Pelargonium species (Geraniaceae).
Lis-Balchin M and Roth G.
J. Essent. Oil Res., 11(1), 83-85 (1999)
M L Hector et al.
World journal of microbiology & biotechnology, 9(5), 562-565 (1993-09-01)
Eighteen non-sibling mutants of Pseudomonas aeruginosa PAO were isolated that were deficient in the utilization of the β-methyl branched acid citronellic acid but not in the utilization of the unbranched n-octanoic acid (Cau mutants). These mutants are also deficient in
Aaron D Gross et al.
Chemico-biological interactions, 263, 1-6 (2016-12-18)
An outbreak of the southern cattle tick, Rhipicephalus (Boophilus) microplus, (Canestrini), in the United States would have devastating consequences on the cattle industry. Tick populations have developed resistance to current acaricides, highlighting the need to identify new biochemical targets along

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service