Skip to Content
Merck
All Photos(3)

Documents

302546

Sigma-Aldrich

Methyl 2-(bromomethyl)acrylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH2Br)CO2CH3
CAS Number:
Molecular Weight:
179.01
Beilstein:
1852481
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.490 (lit.)

bp

35-37 °C/1.3 mmHg (lit.)

density

1.489 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

COC(=O)C(=C)CBr

InChI

1S/C5H7BrO2/c1-4(3-6)5(7)8-2/h1,3H2,2H3

InChI key

CFTUQSLVERGMHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl (2-bromomethyl)acrylate (MBrMA) may be used as chain transfer agents in the emulsion polymerization of methyl methacrylate (MMA) and styrene. MBrMA can undergo nucleophilic substitution of carboxylic acid to form methyl 2-(acyloxymethyl)acrylates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bunichiro Yamada, Shuzo Aoki, Radical polymerization and copolymerization of methyl 2-(acyloxymethyl)acrylate as hindered 2-substituted acrylate
Kobatake S, et al.
Polymer, 36(2), 413-419 (1995)
Mukund P Sibi et al.
Tetrahedron, asymmetry, 17(4), 516-519 (2006-06-27)
We have investigated the effect of nitrogen protecting groups in radical addition trapping experiments leading to beta(2)-amino acids. Of the three N-protecting groups examined, the phthalimido group was optimal with respect to both yields and enantioselectivity. Additionally, radical additions to
Use of methyl 2-(bromomethyl) acrylate as a chain-transfer agent to yield functionalized macromonomers via conventional and living radical polymerizations.
Bon, SAF, et al.
Macromolecules, 33(16), 5819-5824 (2000)
Thomas Mendgen et al.
Bioorganic & medicinal chemistry letters, 20(19), 5757-5762 (2010-08-24)
The enzyme MurA performs an essential step in peptidoglycan biosynthesis and is therefore a target for the discovery of novel antibacterial compounds. We report here the inhibition of MurA by natural products from tulips (tulipalines and tuliposides), and the structure-activity
Fabrice Denes et al.
The Journal of organic chemistry, 72(2), 398-406 (2007-01-16)
Enantioenriched 3,4-disubstituted beta-prolines have been prepared with a high diastereocontrol through a carbometalation reaction or through a domino Michael addition/carbometalation reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service