Skip to Content
Merck
All Photos(3)

Key Documents

302538

Sigma-Aldrich

3-Cyanobenzoyl chloride

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$100.00
5 G
$284.00

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$100.00
5 G
$284.00

About This Item

Linear Formula:
NCC6H4COCl
CAS Number:
Molecular Weight:
165.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

bp

125-129 °C/11 mmHg (lit.)

mp

42-44 °C (lit.)

functional group

acyl chloride
nitrile

SMILES string

ClC(=O)c1cccc(c1)C#N

InChI

1S/C8H4ClNO/c9-8(11)7-3-1-2-6(4-7)5-10/h1-4H

InChI key

RPESZQVUWMFBEO-UHFFFAOYSA-N

Application

3-Cyanobenzoyl chloride was used in preparation of 3-cyanobenzoyl-glycine methyl ester and 6A,6D-bis[3-(aminomethyl)benzoyl-glycyl-amino]-6A,6D-dideoxy-β-cyclodextrin.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Schaschke et al.
Chemistry & biology, 8(4), 313-327 (2001-04-28)
Human beta-tryptase is a mast cell specific trypsin-like serine protease that is thought to play a key role in the pathogenesis of diverse allergic and inflammatory disorders like asthma and psoriasis. The recently resolved crystal structure revealed that the enzymatically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service