Skip to Content
Merck
All Photos(1)

Key Documents

302112

Sigma-Aldrich

N-Benzyl-L-prolinol

99%

Synonym(s):

(S)-(−)-1-Benzylpyrrolidine-2-methanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$134.00

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$134.00

About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Beilstein:
3663640
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

optical activity

[α]20/D −72.7°, neat

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.541 (lit.)

bp

115-120 °C/0.5 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@@H]1CCCN1Cc2ccccc2

InChI

1S/C12H17NO/c14-10-12-7-4-8-13(12)9-11-5-2-1-3-6-11/h1-3,5-6,12,14H,4,7-10H2/t12-/m0/s1

InChI key

ZAIQBJPTOXDDKA-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor in the synthesis of proline-derived homochiral amine oxides.[1] Starting material for enantiomerically pure 3-hydroxypiperidine, a structural feature present in many natural products.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O'Neil, I.A. et al.
Synlett, 617-617 (1995)
Cossy, J. et al.
Tetrahedron Letters, 36, 549-549 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service