Skip to Content
Merck
All Photos(1)

Key Documents

300993

Sigma-Aldrich

Copper(II) 2-pyrazinecarboxylate

97%

Synonym(s):

2-Pyrazinecarboxylic acid copper(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6CuN4O4
CAS Number:
Molecular Weight:
309.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

SMILES string

O=C(O[Cu]OC(=O)c1cnccn1)c2cnccn2

InChI

1S/2C5H4N2O2.Cu/c2*8-5(9)4-3-6-1-2-7-4;/h2*1-3H,(H,8,9);/q;;+2/p-2

InChI key

CPDQQOYHCXKRBR-UHFFFAOYSA-L

General description

Copper(II) 2-pyrazinecarboxylate is also referred as Cu(II)(2-pyrazinecarboxylate)2.

Application

Copper(II) 2-pyrazinecarboxylate was used in preparation of Ag[Cu(2-pyrazinecarboxylate)2](H2O)(NO3), a Cu(II)Ag(I) mixed-metal coordination polymer. It was also used in synthesis of catena(bis3-iodo)-(μ2-pyrazine)-di-copper (I)).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and crystal structure of the two-dimensional coordination polymer catena (bis (?3-iodo)-(? 2-pyrazine)-di-copper (I)).
Goforth AM, et al.
Journal of Chemical Crystallography, 33(4), 303-306 (2003)
Synthesis and characterization of a novel copper (II)? silver (I) mixed-metal coordination polymer: Ag [Cu (2-pyrazinecarboxylate)< sub> 2</sub>](H< sub> 2</sub> O)(NO< sub> 3</sub>).
Dong Y-B, et al.
Solid State Sciences, 2(3), 335-341 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service