Skip to Content
Merck
All Photos(3)

Key Documents

300977

Sigma-Aldrich

(R)-4-Benzyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Beilstein:
4782551
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

optical activity

[α]18/D +64°, c = 1 in chloroform

mp

88-90 °C

SMILES string

O=C1N[C@@H](CO1)Cc2ccccc2

InChI

1S/C10H11NO2/c12-10-11-9(7-13-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,11,12)/t9-/m1/s1

InChI key

OJOFMLDBXPDXLQ-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-4-Benzyl-2-oxazolidinone may be used as a starting material in the synthesis of enantiopure carbocyclic nucleosides.[1] It may also be used as a chiral auxiliary in the enantioselective synthesis of (2R,2′S)-erythro-methylphenidate.[2]
Used in the synthesis of HIV protease inhibitors.[3]
Versatile chiral auxiliary for asymmetric synthesis. For a recent review, see Aldrichimica Acta.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient asymmetric approach to carbocyclic nucleosides: asymmetric synthesis of 1592U89, a potent inhibitor of HIV reverse transcriptase.
Crimmins MT and King BW.
The Journal of Organic Chemistry, 61, 4192-4193 (1996)
Synlett, 4, 679-683 (2004)
Enantioselective synthesis of (2S, 2' R)-erythro-methylphenidate.
Prashad M, et al.
Tetrahedron Asymmetry, 10(18), 3479-3482 (1999)
Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service