Skip to Content
Merck
All Photos(1)

Key Documents

299383

Sigma-Aldrich

2,2,6-Trimethyl-4H-1,3-dioxin-4-one

technical grade

Synonym(s):

Diketene acetone adduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
2408
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

refractive index

n20/D 1.460 (lit.)

bp

~275 °C (lit.)
65-67 °C/2 mmHg (lit.)

mp

12-13 °C (lit.)

solubility

water: insoluble

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CC1=CC(=O)OC(C)(C)O1

InChI

1S/C7H10O3/c1-5-4-6(8)10-7(2,3)9-5/h4H,1-3H3

InChI key

XFRBXZCBOYNMJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flash pyrolysis of 2,2,6-trimethyl-4H-1,3-dioxin-4-one generates acetylketene. It is a diketene-acetone adduct.

Application

2,2,6-trimethyl-4H-1,3-dioxin-4-one was used for acetoacetylation of aliphatic and aromatic alcohols, amines and thiols. It was also used in preparation of N-alkenyl acetoacetamides.

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2, 2, 6-trimethyl-4H-1, 3-dioxin-4-one with imines: an easy route to enamides.
D'Annibale A, et al.
Tetrahedron Letters, 37(41), 7429-7432 (1996)
Acetoacetylation with 2, 2, 6-trimethyl-4H-1, 3-dioxin-4-one: a convenient alternative to diketene.
Clemens RJ and Hyatt JA.
The Journal of Organic Chemistry, 50(14), 2431-2435 (1985)
David M. Birney et al.
The Journal of organic chemistry, 62(21), 7114-7120 (2001-10-24)
Acetylketene (1) was generated by flash pyrolysis of 2,2,6-trimethyl-4H-1,3-dioxin-4-one (6). The selectivities of 1 toward a number of representative functional groups were measured for the first time in a series of competitive trapping reactions. The trend in reactivities toward 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service