Skip to Content
Merck
All Photos(1)

Key Documents

298441

Sigma-Aldrich

Chloroborane methyl sulfide complex

Synonym(s):

Boron monochloride methyl sulfide complex, Chloro(dimethyl sulfide)dihydroboron, Monochloroborane-dimethyl sulfide, monoChloroborane methyl sulfide complex

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$538.00

$538.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$538.00

About This Item

Linear Formula:
(CH3)2S·BH2Cl
CAS Number:
Molecular Weight:
110.41
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$538.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reagent type: reductant

density

1.059 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

C[S+](C)[BH2-]Cl

InChI

1S/C2H8BClS/c1-5(2)3-4/h3H2,1-2H3

InChI key

CUKLQTNDSCOWTI-UHFFFAOYSA-N

Application

Reactant for:
  • Substitution reactions
  • Boronation reactions
  • Preparation of diborylamines
  • Preparation of unsymmetrical borafluorenes via low-temperature carbon-hydrogen activation reactions
  • Asymmetrical aldol reactions with a-chiral aldehydes
  • Addition reaction with α-pinene
Reagent for hydroboration and synthesis of chlorodialkylboranes.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service