Skip to Content
Merck
All Photos(2)

Key Documents

297003

Sigma-Aldrich

2-Amino-3-nitrophenol

98%

Synonym(s):

2-Hydroxy-6-nitroaniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$44.20

$44.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$44.20

About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
2804307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$44.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

212-213 °C (lit.)

functional group

nitro

SMILES string

Nc1c(O)cccc1[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-6-4(8(10)11)2-1-3-5(6)9/h1-3,9H,7H2

InChI key

KUCWUAFNGCMZDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-3-nitrophenol has been used:
  • as semipermanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products[1]
  • as matrix during visible matrix-assisted laser desorption/ionization (VIS-MALDI) mass spectrometry[2]
  • in preparation of 1,3-benzoxazole-4,7-dione[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective preparation of 5-amino-and 6-amino-1, 3-benzoxazole-4, 7-diones from symmetrical diaminophenol and aminoresorcinol.
Brehu L, et al.
Tetrahedron Letters, 46(9), 1437-1440 (2005)
Christina L Burnett et al.
International journal of toxicology, 28(6 Suppl 2), 217S-251S (2010-01-30)
2-Amino-3-nitrophenol, 2-amino-4-nitrophenol, 2-amino-5-nitrophenol, 4-amino-3-nitrophenol, 4-amino-2-nitrophenol, 2-amino-4-nitrophenol sulfate, 3-nitro-p-hydroxyethylaminophenol, and 4-hydroxypropylamino-3-nitrophenol are substituted aromatic compounds used as semipermanent (nonoxidative) hair colorants and as toners in permanent (oxidative) hair dye products. All ingredients in this group except 2-amino-4-nitrophenol sulfate, 2-amino-5-nitrophenol, and 4-amino-2-nitrophenol
Lee Chuin Chen et al.
Rapid communications in mass spectrometry : RCM, 21(24), 4129-4134 (2007-11-21)
Visible matrix-assisted laser desorption/ionization (VIS-MALDI) was performed using 2-amino-3-nitrophenol as matrix. The matrix is of near-neutral pH, and has an optical absorption band in the near-UV and visible region. A frequency-doubled Nd:YAG laser operated at 532 nm wavelength was used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service