Skip to Content
Merck
All Photos(1)

Key Documents

296279

Sigma-Aldrich

2,4-Dimethoxy-3-methylbenzaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
99%

Assay

99%

form

solid

mp

52-54 °C (lit.)

SMILES string

COc1ccc(C=O)c(OC)c1C

InChI

1S/C10H12O3/c1-7-9(12-2)5-4-8(6-11)10(7)13-3/h4-6H,1-3H3

InChI key

UOKAZUWUHOBBMD-UHFFFAOYSA-N

Application

2,4-Dimethoxy-3-methylbenzaldehyde has been used:
  • as starting reagent in stereocontrolled total synthesis of (−)-kendomycin[1]
  • in total syntheses of renierol, renierol acetate and renierol propionate[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New regioselective total syntheses of antibiotic renierol, renierol acetate, and renierol propionate from the 5-oxygenated isoquinoline.
Kuwabara N, et al.
Chemical & Pharmaceutical Bulletin, 47(12), 1805-1807 (1999)
Amos B Smith et al.
Journal of the American Chemical Society, 128(15), 5292-5299 (2006-04-13)
A convergent stereocontrolled total synthesis of (-)-kendomycin (1) has been achieved. The synthesis proceeds with a longest linear sequence of 21 steps, beginning with commercially available 2,4-dimethoxy-3-methylbenzaldehyde (12). Highlights of the synthesis include an effective Petasis-Ferrier union/rearrangement tactic to construct

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service