Skip to Content
Merck
All Photos(1)

Key Documents

295523

Sigma-Aldrich

Methyl vinyl ether

≥98%

Synonym(s):

Methoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH=CH2
CAS Number:
Molecular Weight:
58.08
Beilstein:
1209255
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥98%

contains

0.1% triethanolamine as stabilizer

SMILES string

COC=C

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in lecture bottle

Recommended products

Stainless steel control valve Z146951 or either of stainless steel regulators Z148504 and Z148512 is recommended.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-76.0 °F - closed cup

Flash Point(C)

-60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service