Skip to Content
Merck
All Photos(1)

Key Documents

29550

Sigma-Aldrich

N-Cyclohexylsulfamic acid

≥98.0% (T)

Synonym(s):

Cyclamic acid, Cyclohexanesulfamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$360.00

$360.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$360.00

About This Item

Empirical Formula (Hill Notation):
C6H13NO3S
CAS Number:
Molecular Weight:
179.24
Beilstein:
2208885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$360.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (T)

form

solid

mp

~180 °C (dec.)

solubility

dioxane: 1 g/10 mL, clear, colorless (hot)

SMILES string

OS(=O)(=O)NC1CCCCC1

InChI

1S/C6H13NO3S/c8-11(9,10)7-6-4-2-1-3-5-6/h6-7H,1-5H2,(H,8,9,10)

InChI key

HCAJEUSONLESMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The N-cyclohexylsulfamic acid salts of well−known therapeutic agents were prepared[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of physical properties of certain antitussive and antihistaminic agents by formation of N-cyclohexylsulfamate salts.
J A CAMPBELL et al.
Journal of pharmaceutical sciences, 51, 931-934 (1962-10-01)
Lin Yi et al.
Analytical and bioanalytical chemistry, 386(3), 666-674 (2006-05-26)
Characterizing the biological effects of metabolic transformations (or biotransformation) is one of the key steps in developing safe and effective pharmaceuticals. Sulfate conjugation, one of the major phase II biotransformations, is the focus of this study. While this biotransformation typically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service