Skip to Content
Merck
All Photos(1)

Key Documents

295027

Sigma-Aldrich

Boron trifluoride

≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BF3
CAS Number:
Molecular Weight:
67.81
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.38 (21 °C, vs air)

Quality Level

Assay

≥99.5%

form

gas

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst
reaction type: Polymerization Reactions

bp

−100 °C (lit.)

mp

−127 °C (lit.)

SMILES string

FB(F)F

InChI

1S/BF3/c2-1(3)4

InChI key

WTEOIRVLGSZEPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Monel control valve Z146978 or Monel gas regulator Z562483 is recommended.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brieuc Guillerm et al.
Macromolecular rapid communications, 33(19), 1600-1612 (2012-07-05)
Polyoxazolines (POx) are increasingly being studied as polymeric building blocks due to the possibility of affording tunable properties. Additionally, as the biocompatibility and stealth behavior of POx are similar to that of poly(ethylene glycol) (PEG), it has become challenging to
Ahmad Makahleh et al.
Talanta, 81(1-2), 20-24 (2010-03-02)
A reversed-phase high-performance liquid chromatographic method with capacitively coupled contactless conductivity detector (C(4)D) has been developed for the separation and the simultaneous determination of five underivatized long chain fatty acids (FAs), namely myristic, palmitic, stearic, oleic, and linoleic acids. An
Nolan R Mente et al.
The Journal of organic chemistry, 73(20), 7963-7970 (2008-09-18)
The total synthesis of the natural stilbene (+)-schweinfurthin G (8) has been accomplished through a sequence based on an efficient cationic cascade cyclization. This cascade process is initiated by Lewis acid promoted ring opening of an epoxide and terminated through
G K Surya Prakash et al.
Organic letters, 13(15), 4128-4131 (2011-07-14)
The one-pot synthesis of 1,1,1-trifluoro- and 1,1-difluoro-2,2-diarylethanes from arenes and fluorinated hemiacetals in the BF(3)-H(2)O system is described. The reaction is simple, clean, and convenient, eliminating the use of organic solvents and other expensive acid systems. BF(3)-H(2)O is economic, is
Gewu Lu et al.
ACS nano, 2(11), 2342-2348 (2009-02-12)
Polythiophene (Pth) films with aligned nanotubule arrays were adhered strongly on various smooth surfaces such as glass, mica, and GaAs after drying from their wet states under ambient condition. The normal and shear dry adhesion forces of the films on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service