Skip to Content
Merck
All Photos(1)

Key Documents

293946

Sigma-Aldrich

Octadecyl sulfate sodium salt

technical grade, 93%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$137.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$137.00

About This Item

Linear Formula:
CH3(CH2)17OSO3Na
CAS Number:
Molecular Weight:
372.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

93%

mp

211-213 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C18H38O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-23(19,20)21;/h2-18H2,1H3,(H,19,20,21);/q;+1/p-1

InChI key

NWZBFJYXRGSRGD-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties of ionized monolayers. Part 5.-Surface potentials and pressures of insoluble monolayers of sodium octadecyl sulphate.
Mingins J and Pethica BA.
Transactions of the Faraday Society, 59, 1892-1905 (1963)
Nan Zhang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 116, 66-75 (2016-12-19)
Nanoparticles can improve topical drug delivery: size, surface properties and flexibility of polymer nanoparticles are defining its interaction with the skin. Only few studies have explored skin penetration for one series of structurally related polymer particles with systematic alteration of
Concepción Abril et al.
Chemosphere, 268, 128823-128823 (2020-11-09)
The uptake, bioconcentration and translocation of 22 endocrine disrupting compounds (six perflurocarboxylic acids (PFAAs), perfluorooctanoic sulfonic acid, four anionic surfactants (alkylsulfates (ASC) from C12 to C16), bisphenol A (BPA), four preservatives (parabens), two biocides (triclosan (TCS) and triclocarban (TCB)) and
Astrid Von Mentzer et al.
Virulence, 11(1), 381-390 (2020-04-05)
The ability to adhere via colonization factors to specific receptors located on the intestinal mucosa is a key virulence factor in enterotoxigenic Escherichia coli (ETEC) pathogenesis. Here, the potential glycosphingolipid receptors of the novel human ETEC colonization factor CS30 were
Concepción Abril et al.
The Science of the total environment, 711, 135048-135048 (2019-12-10)
In this work, twenty-three endocrine disrupting compounds have been monitored in sludge from different stages of four sludge stabilization treatments (anaerobic digestion, aerobic digestion, composting and anaerobic stabilization ponds). Their occurrence and fate in sludge stabilization plants and their potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service