Skip to Content
Merck
All Photos(1)

Key Documents

293369

Sigma-Aldrich

3-Aminopiperidine dihydrochloride

97%

Synonym(s):

3-Piperidinamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2 · 2HCl
Molecular Weight:
173.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

230 °C (dec.) (lit.)

SMILES string

Cl.Cl.NC1CCCNC1

InChI

1S/C5H12N2.2ClH/c6-5-2-1-3-7-4-5;;/h5,7H,1-4,6H2;2*1H

InChI key

GGPNYXIOFZLNKW-UHFFFAOYSA-N

General description

Estimation of (S)-piperidin-3-amine in (R)-piperidin-3-amine dihydrochloride (3-aminopiperidine dihydrochloride) has been investigated by chiral high-performance liquid chromatographic (chiral HPLC) method[1].

Application

Reactant for N-arylation of heterocyclic diamines

Reactant for synthesis of substituted quinolones with reduced phototoxic risk

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shubo Dong et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 33(3), 293-298 (2017-03-18)
A sensitive non-derivatization method for the determination of the highly polar compound 3-aminopiperidine was developed using a mixed-mode column combined with a charged aerosol detector (CAD). Chromatographic conditions, including the type of detector, separation mode, and mobile phase composition, were
Chittabathina Venkata Raghunadha Babu et al.
Chirality, 26(12), 775-779 (2014-07-22)
A simple, precise, accurate, robust chiral high-performance liquid chromatographic (chiral HPLC) method was developed for estimation of (S)-piperidin-3-amine (S-isomer) in (R)-piperidin-3-amine dihydrochloride (R-AMP). As AMP is a high-melting solid and nonchromophoric compound, development of a suitable chiral method is a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service