Skip to Content
Merck
All Photos(1)

Key Documents

292915

Sigma-Aldrich

2-Pyrrolidone-5-carboxylic acid

99%

Synonym(s):

DL-5-Oxo-2-pyrrolidinecarboxylic acid, DL-Pyroglutamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$45.00
100 G
$146.00

$45.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$45.00
100 G
$146.00

About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
82134
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$45.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

reaction suitability

reaction type: solution phase peptide synthesis

mp

183-185 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C1CCC(=O)N1

InChI

1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)

InChI key

ODHCTXKNWHHXJC-UHFFFAOYSA-N

General description

DL-Pyroglutamic acid serves as a chiral building block and a chiral auxiliary in asymmetric synthesis.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyroglutamic acid: a versatile building block in asymmetric synthesis
C Najera, et al.
Tetrahedron Asymmetry, 10, 2245-2303 (1999)
Chien-Ming Hsieh et al.
Pharmaceutics, 12(6) (2020-06-18)
Extracellular matrix (ECM) hydrogel can create a favorable regenerative microenvironment and act as a promising dressing for accelerating the healing of diabetic wound. In this study, a simple and effective decellularization technique was developed and optimized to obtain acellular extracellular
Stefania Perticaroli et al.
Biochimica et biophysica acta. Biomembranes, 1861(2), 403-409 (2018-11-23)
The superficial layer of the skin, the stratum corneum (SC), consists of corneocytes surrounded by lipid regions and acts as a protective barrier for the body against water loss, toxic agents and microorganisms. As most substances permeate the stratum corneum
Andres Gil et al.
Journal of pharmaceutical and biomedical analysis, 160, 289-296 (2018-08-17)
Oxidative stress is suggested to play an important role in several pathophysiological conditions. A recent study showed that decreasing 5-oxoproline (pyroglutamate) concentration, an important mediator of oxidative stress, by over-expressing 5-oxoprolinase, improves cardiac function post-myocardial infarction in mice. The aim
Jessica Fiori et al.
Journal of chromatography. A, 1562, 115-122 (2018-06-04)
Monomeric catechins are important compounds in green tea accounting for potential bioactivity against a wide range of diseases. Besides catechins, l-Theanine (γ-glutamylethylamide), a characteristic amino acid in tea leaves, has become a further focus of the phytochemical research for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service