Skip to Content
Merck
All Photos(1)

Key Documents

290505

Sigma-Aldrich

2-Fluorostyrene

contains 4-tert-butylcatechol as inhibitor, 98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$291.00

$291.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$291.00

About This Item

Linear Formula:
H2C=CHC6H4F
CAS Number:
Molecular Weight:
122.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$291.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

contains

4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.52 (lit.)

bp

29-30 °C/4 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccccc1C=C

InChI

1S/C8H7F/c1-2-7-5-3-4-6-8(7)9/h2-6H,1H2

InChI key

YNQXOOPPJWSXMW-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service