Skip to Content
Merck
All Photos(1)

Key Documents

285528

Deoxycorticosterone acetate

98%

Synonym(s):

11-Deoxycorticosterone acetate, 21-Acetoxy-4-pregnene-3,20-dione, 21-Acetoxyprogesterone, 21-Hydroxy-4-pregnene-3,20-dione 21-acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnen-21-ol-3,20-dione 21-acetate, Cortexone acetate, DOC acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
Beilstein:
2570798
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

form

solid

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@]([H])(CC[C@@]24[H])C(=O)COC(C)=O

InChI

1S/C23H32O4/c1-14(24)27-13-21(26)20-7-6-18-17-5-4-15-12-16(25)8-10-22(15,2)19(17)9-11-23(18,20)3/h12,17-20H,4-11,13H2,1-3H3/t17-,18-,19-,20+,22-,23-/m0/s1

InChI key

VPGRYOFKCNULNK-ACXQXYJUSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saad Lahlou et al.
Fundamental & clinical pharmacology, 17(3), 323-330 (2003-06-14)
The present study investigated the hypotensive responses to intravenous (i.v.) treatment with the essential oil of Alpinia zerumbet (EOAZ) and its main constituent, terpinen-4-ol (Trp-4-ol), in the experimental model of deoxycorticosterone-acetate (DOCA)-salt hypertensive rat. In both DOCA-salt hypertensive and uninephrectomized
J P Davis et al.
Clinical science (London, England : 1979), 81(1), 73-78 (1991-07-01)
1. Hypertension was induced in male Sprague-Dawley rats by left unilateral nephrectomy and deoxycorticosterone acetate--salt administration. After 5 weeks, arterial systolic blood pressure was significantly elevated in these animals (191.5 +/- 6.0 mmHg, mean +/- SD, n = 17) compared
Riccardo Candido et al.
American journal of hypertension, 18(6), 844-850 (2005-06-01)
The aim of the present study was to compare glomerular permeability alterations associated with experimental hypertension models known to have different effects on the circulating renin-angiotensin system (RAS). Five groups, 10 animals each, were studied. One group served as a
Ashok Jadhav et al.
Experimental biology and medicine (Maywood, N.J.), 234(7), 764-778 (2009-05-12)
In hypertension, elevated levels of oxidative/inflammatory mediators including nuclear factor kappaB (NF-kappaB), activating protein (AP-1), c-Jun-NH2-terminal kinase (JNK), and cell-regulatory proteins such as transforming growth factor beta (TGF-beta), trigger the mobilization of extracellular matrix (ECM) leading to fibrosis, hypertrophy and
Björn Friedrich et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 22(5-6), 465-474 (2008-12-18)
Renal fibrosis is characterized by excessive accumulation of extracellular matrix proteins. Recent findings show that transforming growth factor-beta (TGF-beta) induces a rapid but transient expression of early growth response gene-1 (Egr-1) by skin fibroblasts. The present study aims to define

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service