Skip to Content
Merck
All Photos(2)

Documents

285315

Sigma-Aldrich

Cetylpyridinium bromide hydrate

98%

Synonym(s):

1-Hexadecylpyridinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38BrN · xH2O
CAS Number:
Molecular Weight:
384.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

67-71 °C (lit.)

solubility

ethanol: soluble 5%, hazy (Pale beige)

SMILES string

[Br-].[H]O[H].CCCCCCCCCCCCCCCC[n+]1ccccc1

InChI

1S/C21H38N.BrH.H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H;1H2/q+1;;/p-1

InChI key

HVWZDJSNXREVCK-UHFFFAOYSA-M

Application

Cetylpyridinium bromide hydrate was used in the modification of the zeolite.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngbo Choi et al.
Nanotechnology, 31(33), 335103-335103 (2020-05-06)
The design of smart and functional nanocarriers for drug delivery systems that use a variety of organic and inorganic materials has led to the development of nanomedicines with improved therapeutic efficiency and reduced side effects. In this study, a pH-
Cabrera-Lafaurie WA, et al.
Journal of Environmental Engineering (New York, N.Y.), 2(2), 899-906 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service