Skip to Content
Merck
All Photos(2)

Documents

282669

Sigma-Aldrich

Lithium trifluoromethanesulfonate

96%

Synonym(s):

LiTf, Lithium triflate, Trifluoromethanesulfonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Li
CAS Number:
Molecular Weight:
156.01
Beilstein:
4301818
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

>300 °C (lit.)

SMILES string

[Li+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Li/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

MCVFFRWZNYZUIJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium trifluoromethanesulfonate can be used:
  • As an electrolyte salt to determine the thermal stability of graphite anodes in Li-ion batteries.
  • For the synthesis of solid polymer electrolytes with polyethylene oxide and chitosan, having potential applications in solid state batteries.
  • As a source of triflate anion (CF3SO3) to synthesize ionic liquids for further syntheses of conducting polymer electrolytes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion conducting behaviour of polymer electrolytes containing ionic liquids.
Singh B and Sekhon SS
Chemical Physics Letters, 414(1-3), 34-39 (2005)
The influence of lithium salt on the interfacial reactions controlling the thermal stability of graphite anodes.
Andersson AM, et al.
Electrochimica Acta, 47(12), 1885-1898 (2002)
Solid polymer electrolytes based on polyethylene oxide and lithium trifluoro-methane sulfonate (PEO-LiCF3SO3): Ionic conductivity and dielectric relaxation.
Karan NK, et al.
Solid State Ionics, 179(19-20), 689-696 (2008)
FTIR studies of chitosan acetate based polymer electrolytes.
Osman Z and Arof AK
Electrochimica Acta, 48(8), 993-999 (2003)
Guangmin Zhou et al.
Nature communications, 6, 7760-7760 (2015-07-18)
Lithium-sulphur batteries with a high theoretical energy density are regarded as promising energy storage devices for electric vehicles and large-scale electricity storage. However, the low active material utilization, low sulphur loading and poor cycling stability restrict their practical applications. Herein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service