Skip to Content
Merck
All Photos(2)

Documents

282162

Sigma-Aldrich

Isoquinoline

technical grade, 90-92%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90-92%

form

solid

refractive index

n20/D 1.623 (lit.)

bp

242-243 °C (lit.)

mp

26-28 °C (lit.)

density

1.099 g/mL at 25 °C (lit.)

SMILES string

c1ccc2cnccc2c1

InChI

1S/C9H7N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-7H

InChI key

AWJUIBRHMBBTKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The intensity of fluorescence peak of isoquinoline increases with increasing pressure in polymethylmethacrylate and in polyisobutylene polymeric media.

Application

Isoquinoline (isq) has been used:
  • in the preparation of cis-[(dcbH2)2Ru(isq)2](ClO4)2 [dcbH2 = 4,4′-(CO2H)2-2,2′-bipyridine]
  • to investigate the toxicity of three two-ring and five three-ring azaarenes to the green alga Scenedesmus acuminatus and its relationship with molecular structure

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Property-toxicity relationships of azaarenes to the green alga Scenedesmus acuminatus.
Vlaardingen V, et al.
Environmental Toxicology and Chemistry / Setac, 15(11), 2035-2042 (1996)
Sensitization of n-type TiO2 electrode by a novel isoquinoline ruthenium (II) polypyridyl complex.
Garcia CG, et al.
Journal of the Brazilian Chemical Society, 9(1), 13-15 (1998)
High pressure studies of isoquinoline luminescence in polymeric media.
Rollinson AM, et al.
Chemical Physics Letters, 59(3), 559-561 (1978)
Jörg T Binder et al.
Journal of the American Chemical Society, 134(41), 17003-17006 (2012-10-09)
We have developed a nickel-catalyzed method for the asymmetric cross-coupling of secondary electrophiles with secondary nucleophiles, specifically, stereoconvergent Negishi reactions of racemic benzylic bromides with achiral cycloalkylzinc reagents. In contrast to most previous studies of enantioselective Negishi cross-couplings, tridentate pybox
Asymmetric synthesis of isoquinoline alkaloids.
Maria Chrzanowska et al.
Chemical reviews, 104(7), 3341-3370 (2004-07-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service