Skip to Content
Merck
All Photos(1)

Key Documents

281581

Sigma-Aldrich

(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine

96%

Synonym(s):

(R)-1-Amino-2-(methoxymethyl)pyrrolidine, RAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
Beilstein:
4229866
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

96%

optical activity

[α]18/D +79°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.465 (lit.)

bp

42 °C/1.8 mmHg (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

COC[C@H]1CCCN1N

InChI

1S/C6H14N2O/c1-9-5-6-3-2-4-8(6)7/h6H,2-5,7H2,1H3/t6-/m1/s1

InChI key

BWSIKGOGLDNQBZ-ZCFIWIBFSA-N

Application

Reagent for asymmetric synthesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey R Skaar et al.
Nature reviews. Drug discovery, 13(12), 889-903 (2014-11-15)
The clinical successes of proteasome inhibitors for the treatment of cancer have highlighted the therapeutic potential of targeting this protein degradation system. However, proteasome inhibitors prevent the degradation of numerous proteins, which may cause adverse effects. Increased specificity could be
Kyeong-Hyeon Chun et al.
Journal of hypertension, 38(9), 1777-1785 (2020-07-11)
Exercise capacity is often reduced after heart transplantation. We aimed to investigate the association between aortic stiffness and exercise capacity after heart transplantation. We retrospectively analyzed the data of patients who underwent a cardiopulmonary exercise test and central hemodynamic measurements
Andreas Panagopoulos et al.
Trends in cell biology, 30(4), 290-302 (2020-02-12)
The cullin-RING E3 ubiquitin ligase CRL4Cdt2 has emerged as a master regulator of genome stability, which targets key cell cycle proteins for proteolysis during S phase and after DNA damage. Recent advances shed light on how it couples ubiquitination to
Xiao-Yi Zhou et al.
Ultrasound in medicine & biology, 43(7), 1486-1493 (2017-05-02)
Low-intensity pulsed ultrasound (LIPUS) has been found to accelerate fracture healing. In this study, we analyzed the role of calcitonin gene-related peptide (CGRP) in a rat spinal fusion model treated with LIPUS. The results revealed that LIPUS significantly increases bone
Majid Masso
Journal of theoretical biology, 453, 58-67 (2018-05-22)
Scientific breakthroughs in recent decades have uncovered the capability of RNA molecules to fulfill a wide array of structural, functional, and regulatory roles in living cells, leading to a concomitantly significant increase in both the number and diversity of experimentally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service