Skip to Content
Merck
All Photos(2)

Key Documents

280895

Sigma-Aldrich

4-Bromobenzenediazonium tetrafluoroborate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4N2BF4
CAS Number:
Molecular Weight:
270.82
Beilstein:
3791621
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

135-140 °C (dec.) (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.Brc1ccc(cc1)[N+]#N

InChI

1S/C6H4BrN2.BF4/c7-5-1-3-6(9-8)4-2-5;2-1(3,4)5/h1-4H;/q+1;-1

InChI key

FXTCQUCYDJZGGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical reduction of 4-bromobenzenediazonium tetrafluoroborate was investigated for the derivatization of the carbon felt (modified carbon surfaces whether glassy carbon or carbon felt)[1].

Application

4-Bromobenzenediazonium tetrafluoroborate was used in preparation of:
  • surface modified Au(111) electrode[2]
  • 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical attachment of organic groups to carbon felt surfaces.
Coulon E, et al.
Langmuir, 17(22), 7102-7106 (2001)
Bo Cui et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 2955-2960 (2013-02-16)
Surface modifications of a Au(111) electrode with 4-bromobenzenediazonium tetrafluoroborate (BBD) in acetonitrile (ACN) and 0.1 M HClO4 have been characterized by scanning tunneling microscopy (STM). In ACN, STM results reveal the formation of disordered thin organic films. The involvement of
Efficient access to perfluoroalkylated aryl compounds by Heck reaction.
Darses S, et al.
European Journal of Organic Chemistry, 2001(6), 1121-1128 (2001)
Tomonari Shiraishi et al.
Nanoscale, 9(43), 16900-16907 (2017-10-28)
Single-walled carbon nanotubes (SWNTs) with local chemical modification have been recognized as a novel near infrared (NIR) photoluminescent nanomaterial due to the emergence of a new red-shifted photoluminescence (PL) with enhanced quantum yields. As a characteristic feature of the locally
Younghee Park et al.
ACS applied materials & interfaces, 12(36), 40870-40878 (2020-08-19)
Although surface engineering has been regarded to be a great approach to modulate the optical and electrical properties of nanomaterials, the spontaneous covalent functionalization on semiconducting 2H-MoS2 is a notoriously difficult process, while several reactions have been performed on metallic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service