Skip to Content
Merck
All Photos(1)

Key Documents

280240

Sigma-Aldrich

5-Amino-3,4-dimethylisoxazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2O
CAS Number:
Molecular Weight:
112.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

118-122 °C (lit.)

SMILES string

Cc1noc(N)c1C

InChI

1S/C5H8N2O/c1-3-4(2)7-8-5(3)6/h6H2,1-2H3

InChI key

PYNDWPFZDQONDV-UHFFFAOYSA-N

General description

5-amino-3,4-dimethylisoxazole formed complexes with cobalt(II) and copper(II). The kinetics of gas-phase thermal isomerization of 5-amino-3,4-dimethylisoxazole was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase thermal isomerization of some aminomethylisoxazoles.
Perez JD, et al.
The Journal of Organic Chemistry, 46(17), 3505-3508 (1981)
Synthesis and characterization of cobalt (II) and copper (II) complexes with 5-amino-3, 4-dimethylisoxazole.
Biddau M, et al.
Transition Met. Chem. (London), 2(1), 5-9 (1977)
S M Sicardi et al.
Mutation research, 343(1), 61-66 (1995-05-01)
Studies were undertaken to evaluate the ability of various quinoneimines to induce micronuclei in bone marrow cells as a measure of their genotoxicity. Accordingly, 2-hydroxy-N-(3,4-dimethyl-5-isoxazolyl)-1,4-naphthoquinone-4-imine (I), its 2-acetyl derivative (II) and 2-[(5-methyl-3-isoxazolyl)amino]-N-(5-methyl-3-isoxazolyl)-1 ,4- naphthoquinone-4-imine (III), as well as two of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service