280216
Nα-Acetylglycinamide
97%
Synonym(s):
2-Acetamidoacetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
mp
140-143 °C (lit.)
SMILES string
CC(=O)NCC(N)=O
InChI
1S/C4H8N2O2/c1-3(7)6-2-4(5)8/h2H2,1H3,(H2,5,8)(H,6,7)
InChI key
WQELDIQOHGAHEM-UHFFFAOYSA-N
Application
Nα-Acetylglycinamide (2-Acetamidoacetamide) was used in the preparation of 3-unsubstituted 2-pyridones by a tandem reaction with chalcones.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biulleten' eksperimental'noi biologii i meditsiny, 107(6), 711-713 (1989-06-01)
The effect of electroshock (ECS) and piracetam, oxiracetam or N-acetylglycinamide on the passive avoidance conditioned response in rats was studied. The antiemetic effect of the compounds was examined in cats as well. The results obtained allowed us to distinct the
Biochemistry, 43(5), 1329-1342 (2004-02-06)
With knowledge of individual transfer free energies of chemical groups that become newly exposed on protein denaturation and assuming the group transfer free energy contributions are additive, it should be possible to predict the stability of a protein in the
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 32(5), 844-850 (2012-01-19)
Limited evidence exists on the relationships between severity of white-matter lesions (WMLs) and cerebral hemodynamics in patients without major cerebral artery disease. To examine changes of cerebral blood flow (CBF), oxygen metabolism, and vascular reserve capacity associated with severity of
[The systems analysis of the effect of piracetam, glycine and N-acetylglycinamide on long-term learning].
Eksperimental'naia i klinicheskaia farmakologiia, 56(4), 55-57 (1993-07-01)
A Regioselective Tandem Reaction between Chalcones and 2-Acetamidoacetamide Promoted by Cs2 CO3 for the Preparation of 3-Unsubstituted 2-Pyridones.
Synthesis, 2003(04), 0487-0490 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service