Skip to Content
Merck
All Photos(1)

Key Documents

279927

Sigma-Aldrich

(1R,5S)-(+)-2-Oxabicyclo[3.3.0]oct-6-en-3-one

99%

Synonym(s):

(1R,5S)-2-Oxabicyclo[3.3.0]oct-6-en-3-one, (3aS,6aR)-3,3a,6,6a-Tetrahydro-2H-1-oxapentalen-2-one, (3aS,6aR)-3,3a,6,6a-Tetrahydro-2H-cyclopenta[b]furan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352103

Assay

99%

optical activity

[α]20/D +102.5°, c = 1 in methanol

mp

45-48 °C (lit.)

SMILES string

O=C1C[C@H]2C=CC[C@H]2O1

Application

Building block for the synthesis of carbocyclic nucleosides.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akella, L.B. Vince, R.
Tetrahedron, 52, 8407-8407 (1996)
Hagai Tavori et al.
Bioorganic & medicinal chemistry, 16(15), 7504-7509 (2008-06-24)
Paraoxonase1 (PON1) is a HDL bound enzyme and many of the anti-atherogenic properties of HDL are attributed to PON1. The enzyme precise mechanism of protective action and its endogenous substrate remain elusive. PON1 hydrolyzes organophosphates, arylesters and lactones, whereas the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service