Skip to Content
Merck
All Photos(1)

Key Documents

278882

Sigma-Aldrich

1,1-Dimethyl-2-selenourea

97%

Synonym(s):

1,1-Dimethylselenourea, N,N-Dimethylselenourea

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$127.00

About This Item

Linear Formula:
(CH3)2NC(Se)NH2
CAS Number:
Molecular Weight:
151.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

172-174 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CN(C)C(N)=[Se]

InChI

1S/C3H8N2Se/c1-5(2)3(4)6/h1-2H3,(H2,4,6)

InChI key

NRTHBQJARHPPAU-UHFFFAOYSA-N

General description

One-electron oxidation of 1,1-dimethyl-2-selenourea (1,1′-dimethyl-2-selenourea) by hydroxyl radicals, one-electron-specific oxidants, was studied using pulse radiolysis technique[1].

Application

1,1-Dimethyl-2-selenourea was used as precursor to synthesize CdSe quantum dots in aqueous solution under simultaneous illumination with a diode-pumped solid state green laser and a Xe-Hg lamp[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pulse and ?-radiolysis studies on aqueous solution of 1, 1'-dimethyl-2-selenourea.
Mishra BB, et al.
Radiation Physics and Chemistry, 77(2), 125-130 (2008)
Yang-Wei Lin et al.
Journal of nanoscience and nanotechnology, 6(4), 1092-1100 (2006-06-02)
This paper describes the synthesis of highly water-soluble and fluorescent core-shell-shell silica-CdSe/CdS-silica nanoparticles (CSS silica-QDs-silica NPs). We used cadmium nitrate and 1,1-dimethyl-2-selenourea precursors to synthesize CdSe quantum dots (QDs) in aqueous solution under simultaneous illumination with a diode-pumped solid state

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service