Skip to Content
Merck
All Photos(2)

Key Documents

277789

Sigma-Aldrich

1-Bromo-3,5-dichlorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3Cl2
CAS Number:
Molecular Weight:
225.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

bp

232 °C/757 mmHg (lit.)

mp

73-75 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1cc(Cl)cc(Br)c1

InChI

1S/C6H3BrCl2/c7-4-1-5(8)3-6(9)2-4/h1-3H

InChI key

DZHFFMWJXJBBRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-3,5-dichlorobenzene on treatment with iPrMgCl.LiCl in THF and then DMF at 0°C, followed by the reaction with molecular iodine and aq NH3 yields the corresponding aromatic nitrile[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical one-pot conversion of aryl bromides and ?-bromostyrenes into aromatic nitriles and cinnamonitriles.
Ishii G, et al.
Tetrahedron, 69(5), 1462-1469 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service