All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3C6H4CH2OH
CAS Number:
Molecular Weight:
164.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
refractive index
n20/D 1.515 (lit.)
density
0.962 g/mL at 25 °C (lit.)
SMILES string
CCCCc1ccc(CO)cc1
InChI
1S/C11H16O/c1-2-3-4-10-5-7-11(9-12)8-6-10/h5-8,12H,2-4,9H2,1H3
InChI key
CIDZDYAPPNMBIY-UHFFFAOYSA-N
Application
4-Butylbenzyl alcohol has been used in the preparation of substituted benzyl-pyrimidines.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cécile Gasse et al.
Bioorganic & medicinal chemistry, 16(11), 6075-6085 (2008-05-10)
A series of N(1)-(4-substituted-benzyl)-pyrimidines were synthesized as potential inhibitors of thymidine monophosphate kinase of Mycobacterium tuberculosis (TMPKmt). Key SAR parameters included the chain length substitution in para position of the benzyl ring, the functional group terminating the alkyl chain, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service