Skip to Content
Merck
All Photos(1)

Key Documents

276766

Sigma-Aldrich

1-Acetonaphthone

97%

Synonym(s):

1-Acetylnaphthalene, Methyl 1-naphthyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$62.00

$62.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$62.00

About This Item

Linear Formula:
C10H7COCH3
CAS Number:
Molecular Weight:
170.21
Beilstein:
1100618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.628 (lit.)

bp

302 °C (lit.)

mp

10.5 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cccc2ccccc12

InChI

1S/C12H10O/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-8H,1H3

InChI key

QQLIGMASAVJVON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Stereoselective reduction of 1-acetonapthone in aqueous media by whole cells of Geotrichum candidum has been investigated in various organic co-solvents[1]. Magnetic field effects on the photoinduced electron-transfer reaction between 1-acetonaphthone and diphenylamine in micellar solution have been studied by laser flash photolysis[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic field effects on the photoinduced electron-transfer reaction of 1-acetonaphthone and diphenylamine in micellar solution.
Tanimoto Y, et al.
Chemical Physics Letters, 129(4), 414-418 (1986)
Mani Shankar Bhattacharyya et al.
New biotechnology, 29(3), 359-364 (2011-08-13)
Sixteen organic co-solvents were screened for stereoselective reduction of 1-acetonapthone in aqueous media by whole cells of Geotrichum candidum. Benzyl alcohol was found to be a good co-solvent as it afforded a high coversion and reduced deactivation of the cells.
Babu Balaji et al.
European journal of medicinal chemistry, 92, 332-341 (2015-01-13)
Oxovanadium(IV) complexes [VO(Fc-tpy)(acac)](ClO4) (1), [VO(Fc-tpy)(nap-acac)](ClO4) (2), [VO(Fc-tpy)(py-acac)](ClO4) (3) and [VO(Ph-tpy)(py-acac)](ClO4) (4) of 4'-ferrocenyl-2,2':6',2"-terpyridine (Fc-tpy) and 4'-phenyl-2,2':6',2"-terpyridine (Ph-tpy) having monoanionic acetylacetonate (acac), naphthylacetylacetonate (nap-acac) or pyrenylacetylacetonate (py-acac) ligand were prepared, characterized and their photocytotoxicity in visible light studied. The ferrocenyl complexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service