Skip to Content
Merck
All Photos(1)

Key Documents

275891

Sigma-Aldrich

Dichloro(1,5-cyclooctadiene)palladium(II)

99%

Synonym(s):

PdCl2(cod)

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$325.00

$325.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
2 G
$325.00

About This Item

Empirical Formula (Hill Notation):
C8H12Cl2Pd
CAS Number:
Molecular Weight:
285.51
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

$325.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

99%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

210 °C (dec.) (lit.)

SMILES string

Cl[Pd]Cl.C1CC=CCCC=C1

InChI

1S/C8H12.2ClH.Pd/c1-2-4-6-8-7-5-3-1;;;/h1-2,7-8H,3-6H2;2*1H;/q;;;+2/p-2/b2-1-,8-7-;;;

InChI key

RRHPTXZOMDSKRS-PHFPKPIQSA-L

Application

Catalyst for C-C bond and C-N formation, used in Heck coupling of alkynes with alkenes , Suzuki cross-coupling of aryl bromides , allylic substitution of oximes with allylic acetate , and methoxycarbonylation of iodobenzene.
Dichloro(1,5-cyclooctadiene)palladium(II) is a commonly used catalyst to protect alcohols as acetals by treating with 2-benzyloxy-1-propene; to synthesize bicyclooctanes from diethyl malonate and in alkene carbonylation.[1][2] It can also be used to catalyze the Heck coupling of nonactivated vinyl phosphates with electron deficient alkenes[3]; the synthesis of benzo[b]thiophenes from thioenols[4] and to prepare an active catalyst for Mizoroki-Heck and Suzuki-Miyaura cross-coupling reactions.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An inexpensive and highly stable ligand 1, 4-bis (2-hydroxy-3, 5-di-tert-butylbenzyl) piperazine for Mizoroki?Heck and room temperature Suzuki?Miyaura cross-coupling reactions.
Mohanty S, et al.
Tetrahedron, 64(1), 240-247 (2008)
Cyclic Diolefin Complexes of Platinum and Palladium.
Inorg. Synth. (2007)
Palladium-catalysed direct synthesis of benzo [b] thiophenes from thioenols.
Inamoto K, et al.
Chemical Communications (Cambridge, England), 43, 5529-5531 (2008)
Studies on the Heck reaction with alkenyl phosphates: Can the 1, 2-migration be controlled? Scope and Limitations.
Ebran J P, et al.
Journal of the American Chemical Society, 129(21), 6931-6942 (2007)
Dichloro(1,5?cyclooctadiene)palladium(II).
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service