Skip to Content
Merck
All Photos(1)

Key Documents

274763

Sigma-Aldrich

DL-p-Chloroamphetamine hydrochloride

98%

Synonym(s):

4-Chloro-α-methylphenethylamine hydrochloride, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CH(CH3)NH2·HCl
CAS Number:
Molecular Weight:
206.11
MDL number:
UNSPSC Code:
12352103

Assay

98%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

168-170 °C (lit.)

SMILES string

Cl.CC(N)Cc1ccc(Cl)cc1

InChI

1S/C9H12ClN.ClH/c1-7(11)6-8-2-4-9(10)5-3-8;/h2-5,7H,6,11H2,1H3;1H

InChI key

DZAANUYJOGCNLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service