Skip to Content
Merck
All Photos(1)

Key Documents

27455

Sigma-Aldrich

(±)-Potassium citramalate monohydrate

≥95.0% (NT)

Synonym(s):

(±)-2-Hydroxy-2-methylsuccinic acid dipotassium salt, (±)-2-Methylmalic acid dipotassium salt, (±)-Citramalic acid dipotassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$185.00

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$185.00

About This Item

Linear Formula:
KOOCCH2C(OH)(CH3)COOK · H2O
CAS Number:
Molecular Weight:
242.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (NT)

form

solid

SMILES string

O.[K+].[K+].CC(O)(CC([O-])=O)C([O-])=O

InChI

1S/C5H8O5.2K.H2O/c1-5(10,4(8)9)2-3(6)7;;;/h10H,2H2,1H3,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2

InChI key

QYWXEUYISHLCOP-UHFFFAOYSA-L

Related Categories

Application

Potassium citramalate monohydrate may be used in chemical synthesis studies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metodi Dimitrov Metodiev et al.
Journal of medical genetics, 51(12), 834-838 (2014-10-30)
Inherited optic neuropathy has been ascribed to mutations in mitochondrial fusion/fission dynamics genes, nuclear and mitochondrial DNA-encoded respiratory enzyme genes or nuclear genes of poorly known mitochondrial function. However, the disease causing gene remains unknown in many families. We used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service