Skip to Content
Merck
All Photos(1)

Key Documents

273953

Sigma-Aldrich

4-(3,4-Dimethoxyphenyl)butyric acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$149.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$149.00

About This Item

Linear Formula:
(CH3O)2C6H3(CH2)3CO2H
CAS Number:
Molecular Weight:
224.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

bp

210 °C/6 mmHg (lit.)

mp

62-63 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccc(CCCC(O)=O)cc1OC

InChI

1S/C12H16O4/c1-15-10-7-6-9(8-11(10)16-2)4-3-5-12(13)14/h6-8H,3-5H2,1-2H3,(H,13,14)

InChI key

IDQKCIAJGRMMNC-UHFFFAOYSA-N

Application

4-(3,4-Dimethoxyphenyl)butyric acid may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service