Skip to Content
Merck
All Photos(1)

Key Documents

273457

Sigma-Aldrich

N-Guanylurea sulfate salt hydrate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$233.00

$233.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$233.00

About This Item

Linear Formula:
[H2NC(=NH)NHCONH2]2·H2SO4 · xH2O
CAS Number:
Molecular Weight:
151.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$233.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

199 °C (dec.) (lit.)

functional group

amine

SMILES string

O.OS(O)(=O)=O.NC(=N)NC(N)=O.NC(=N)NC(N)=O

InChI

1S/2C2H6N4O.H2O4S.H2O/c2*3-1(4)6-2(5)7;1-5(2,3)4;/h2*(H6,3,4,5,6,7);(H2,1,2,3,4);1H2

InChI key

DZDHHDIWWOAIGP-UHFFFAOYSA-N

Application

N-Guanylurea sulfate salt hydrate may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baptiste A J Poursat et al.
Ecotoxicology and environmental safety, 182, 109414-109414 (2019-07-14)
Metformin (MET) is a pharmaceutical product mostly biotransformed in the environment to a transformation product, guanylurea (GUA). In ready biodegradability tests (RBTs), however, contrasting results have been observed for metformin. The objective of this study was to measure the biodegradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service