Skip to Content
Merck
All Photos(1)

Key Documents

273252

Sigma-Aldrich

3,3-Diethoxy-1-propanol

98%

Synonym(s):

3,3-Diethoxy-1-propanol, 3,3-Diethoxypropan-1-ol, 3,3-Diethoxypropanol, 3-Hydroxypropanal diethyl acetal, 3-Hydroxypropionaldehyde diethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$74.10
5 G
$229.00

$74.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$74.10
5 G
$229.00

About This Item

Linear Formula:
(C2H5O)2CHCH2CH2OH
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

90-93 °C/14 mmHg (lit.)

density

0.941 g/mL at 25 °C (lit.)

SMILES string

CCOC(CCO)OCC

InChI

1S/C7H16O3/c1-3-9-7(5-6-8)10-4-2/h7-8H,3-6H2,1-2H3

InChI key

ASERXEZXVIJBRO-UHFFFAOYSA-N

Application

3,3-Diethoxy-1-propanol was used in the synthesis of α-acetal-poly(ethylene glycol) via reaction with potassium napthalide and ethylene oxide inside an UniLab Glovebox[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirk Grafahrend et al.
Biotechnology and bioengineering, 101(3), 609-621 (2008-05-08)
Electrospun fibers that are protein resistant and functionalized with bioactive signals were produced by solution electrospinning amphiphilic block copolymers. Poly (ethylene glycol)-block-poly(D,L-lactide) (PEG-b-PDLLA) was synthesized in two steps, with a PEG segment of 10 kDa, while the PDLLA block ranged
Faezeh Ghorbanizamani et al.
Archives of biochemistry and biophysics, 695, 108643-108643 (2020-10-31)
Artificial nanostructures using polymers to produce polymeric vesicles are inspired by the many intricate structures found in living organisms. Polymersomes are a class of self-assembled vesicles known for their great stability and application in drug delivery. They can be tuned
Babita Shashni et al.
Biomaterials, 134, 143-153 (2017-05-02)
Early diagnosis of metastatic cancers could greatly limit the number of cancer-associated deaths. Aberrant surface expression of sialic acid (hypersialylation) on tumors correlating with metastatic incidence and its involvement in tumorigenesis and progression is widely reported; hence detection of hypersialylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service