Skip to Content
Merck
All Photos(2)

Key Documents

271969

Sigma-Aldrich

Poly(vinyl sulfate) potassium salt

average Mw ~170,000

Synonym(s):

potassium ethenyl sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$96.90
5 G
$339.00

$96.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$96.90
5 G
$339.00

About This Item

Linear Formula:
[CH2CH(OSO3K)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$96.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mw ~170,000

SMILES string

[K]OS(=O)(=O)OC=C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Hattori et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(4), 543-546 (2008-04-12)
Polyion complexes of three chitosans with poly(vinyl sulfate) (PVS) and dodecylbenzene sulfonate (DBS) were examined by a potentiometric study that was to separately measure the pH of sample solutions individually prepared. Apparent formation constants (Ki) of ion association between the
T T Luong et al.
European journal of pharmacology, 208(3), 213-221 (1991-11-13)
Inositol 1,4,5-trisphosphate (InsP3) serves as a second messenger for Ca2+ mobilization in a wide variety of cells. InsP3 activates a specific receptor/channel located on an internal Ca2+ store. Because heparin has already been shown to block the action of InsP3
Yasuo Yoshioka et al.
Biochimica et biophysica acta, 1624(1-3), 54-59 (2003-12-04)
We previously reported the development of a "cytomedicine" that consists of cells trapped in alginate-poly-L-lysine-alginate (APA) microcapsules and agarose microbeads. The functional cells that are entrapped in semipermeable polymer are completely isolated from cellular immune system. However, the ability of
Takato Mizunuma et al.
Colloids and surfaces. B, Biointerfaces, 56(1-2), 155-160 (2006-12-22)
We studied the immobilization of a mycelium (Aspergillus niger) using the working hypothesis as follows: (a) when polycation was added to the cell suspension, a few parts of it would bind on the surface of a hypha, allowing to gather
Nirmal Prabhakar et al.
Analytica chimica acta, 589(1), 6-13 (2007-04-03)
Double stranded calf thymus deoxyribonucleic acid entrapped polypyrrole-polyvinyl sulphonate (dsCT-DNA-PPy-PVS) films fabricated onto indium-tin-oxide (ITO) coated glass plates have been used to detect organophosphates such as chlorpyrifos and malathion. These disposable dsCT-DNA-PPy-PVS/ITO bioelectrodes have been characterized using cyclic voltammetry, Fourier-transform-infra-red

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service