Skip to Content
Merck
All Photos(1)

Key Documents

271403

Sigma-Aldrich

Bis(tributylstannyl)acetylene

95%

Synonym(s):

1,1′-(1,2-Ethynediyl)bis[1,1,1-tributylstannane], Bis(tri-n-butylstannyl)acetylene, Bis(tributylstannyl)ethyne, Bis(tributyltin)acetylene, Ethynylenebis[tributyltin]

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$323.00

$323.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$323.00

About This Item

Linear Formula:
[CH3(CH2)3]3SnC≡CSn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
604.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$323.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.493 (lit.)

density

1.147 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC

InChI

1S/6C4H9.C2.2Sn/c6*1-3-4-2;1-2;;/h6*1,3-4H2,2H3;;;

InChI key

CAUONNQGFXOEMY-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service