Skip to Content
Merck
All Photos(1)

Key Documents

270180

Sigma-Aldrich

3-(Aminomethyl)-PROXYL

Synonym(s):

3-(Aminomethyl)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$221.00

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C9H19N2O
CAS Number:
Molecular Weight:
171.26
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

bp

128-131 °C/13 mmHg (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CC1(C)CC(CN)C(C)(C)N1[O]

InChI

1S/C9H19N2O/c1-8(2)5-7(6-10)9(3,4)11(8)12/h7H,5-6,10H2,1-4H3

InChI key

MMNYKXJVNIIIEG-UHFFFAOYSA-N

Application

Useful spin label for studying biological systems.

Used to study molecular diffusion into horse spleen ferritin and has been shown to be an effective nonthiol radioprotector.

Used in pharmacological studies of myeloperoxidase-mediated hypochlorous acid production inhibition

Reactant involved in:
  • Reduction reactions with nitroxides
  • Synthesis of adenosine methycarboxamides for use as cardioprotectants

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

X Yang et al.
Biophysical journal, 78(4), 2049-2059 (2000-03-29)
The detailed kinetics of permeation and effusion of small nitroxide spin probe radicals with the protein shells of horse spleen ferritin (HoSF) and human H-chain ferritin (HuHF) and a 3-fold channel variant D131H+E134H of HuHF were studied by electron paramagnetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service