Skip to Content
Merck
All Photos(1)

Key Documents

269891

Sigma-Aldrich

Tetrahydrofuran-d8

≥99.5 atom % D, contains 1 % (v/v) TMS

Synonym(s):

THF-d8, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$792.00

$792.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$792.00

About This Item

Empirical Formula (Hill Notation):
C4D8O
CAS Number:
Molecular Weight:
80.16
Beilstein:
111854
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

$792.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

isotopic purity

≥99.5 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

contains

1 % (v/v) TMS

technique(s)

NMR: suitable

refractive index

n20/D 1.403 (lit.)

bp

65-66 °C (lit.)

mp

−106 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

WYURNTSHIVDZCO-SVYQBANQSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Experimental and Theoretical Study of CO2 Insertion into Ruthenium Hydride Complexes.: This research provides insights into the mechanistic pathways for CO2 reactivity with ruthenium hydride complexes, potentially relevant for catalytic applications including transformations involving Tetrahydrofuran-d₈ (Ramakrishnan et al., 2016).


  • Syntheses, structures, and NMR chemical shifts of a family of trimethyltin alkoxide, amide, halide and cyclopentadienyl compounds.: Details the synthesis and structural characterization of trimethyltin compounds, useful for understanding the coordination chemistry that may involve Tetrahydrofuran-d₈ as a solvent or structural analogue (Lichtscheidl et al., 2015).


  • NMR studies of coupled low- and high-barrier hydrogen bonds in pyridoxal-5′-phosphate model systems in polar solution.: Discusses the application of NMR spectroscopy in studying hydrogen bonding interactions, where Tetrahydrofuran-d₈ could be utilized as a deuterated solvent for enhanced spectral clarity (Sharif et al., 2007).


  • Stable hydrocarbon diradical, an analogue of trimethylenemethane.: Investigates stable hydrocarbon diradicals, where Tetrahydrofuran-d₈ may be used as part of the experimental setup to stabilize reactive intermediates or as a solvent to study radical stability (Rajca et al., 2005).



Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Far-Infrared Spectrum of Tetrahydrofuran: Spectroscopic Evidence for Pseudorotation.
Lafferty WJ, et al.
J. Chem. Phys. , 42(8), 2915-2919 (1965)
Dynamic NMR study of the interference between cyclic proton exchange, selfassociation and hindered rotation of diphenylformamidine in tetrahydrofuran.
Meschede L, et al.
Berichte der Bunsengesellschaft fur physikalische Chemie, 92(4), 469-485 (1988)
NMR lineshape theory of superimposed intermolecular spin exchange reactions and its action to the system acetic acid/methanol/tetrahydrofuran-d8.
Limbach HH.
Journal of Magnetic Resonance, 36(3), 287-300 (1979)
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service