Skip to Content
Merck
All Photos(1)

Key Documents

269832

Sigma-Aldrich

Nitromethane-d3

99 atom % D

Synonym(s):

Trideuteronitromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3NO2
CAS Number:
Molecular Weight:
64.06
Beilstein:
1751076
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

≥98.50%

form

liquid

expl. lim.

7.1-63 % (lit.)

packaging

pk of 10 × 1 mL

technique(s)

NMR: suitable

impurities

≤0.1000% water
water

refractive index

n20/D 1.3795 (lit.)

bp

100 °C (lit.)

mp

-29 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3/i1D3

InChI key

LYGJENNIWJXYER-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotope effects on selected physicochemical properties of nitromethane and 1-pentanol.
Makowska A and Szydlowski J.
Nukleonika, 51, 81-85 (2006)
DFT molecular structure and initial assignment for the FTIR spectrum of ?7 band of nitromethane-D3.
Pal C and Kshirsagar RJ.
Indian J. Phys., 84(8), 929-937 (2010)
Pyrolysis of nitromethane-d3.
Crawforth CG and Waddington DJ.
The Journal of Physical Chemistry, 74(4), 2793-2796 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service